Learn More
3M™ Reusable Low Maintenance Half Face Mask Respirator
Reusable Half Face Masks 6000 Series are made from a lightweight, comfortable elastomeric material with a bayonet-style twin-filter design, cradle head harness and easy-fasten neck strap.
Brand: 3M™ 7100049928
171.75 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For economical, low-maintenance reusable masks that are easy to handle, use 3M™ Reusable Half Face Mask 6000 Series.
- Our reusable half masks are designed for simplicity, with an elastomeric material that’s lightweight and comfortable to wear over long shifts, a cradle head harness and easy-fasten neck strap.
- They provide protection against gases, vapours and particulates, and feature a bayonet-style connection that fits to a broad range of twin, lightweight filters, depending on your individual needs.
- The twin-filter design evenly balances weight across the mask, and the low profile means minimal impact to your field of vision and better compatibility with welding and grinding shields.
- They can be used with 3M™ Supplied Air Systems for your increased convenience and flexibility, and connect with all 3M™ 2000, 5000 and 6000 Series filters (except 3M™ 6098 or 6099).
- The 6000 Series has a specially designed range of spares and accessories to help you maintain the right level of comfort and protection when parts become worn or damaged.
- They’re available in three sizes; small, medium and large.
Specifications
Gray | |
Reusable | |
Thermoplastic Elastomer | |
Large | |
Yes |
Bayonet Style | |
Combination | |
Reusable Half Facepiece | |
No |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
CE_17893256_EN.pdf
File size: 880.8 kB
Product Content Correction
Product Title
3M™ Reusable Low Maintenance Half Face Mask Respirator > Size: Large
Spot an opportunity for improvement?Share a Content Correction