Learn More
RHOT1 Monoclonal Antibody (CL1083), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524622
Description
Immunogen sequence: THIVDYSEAE QSDEQLHQEI SQANVICIVY AVNNKHSIDK VTSRWIPLIN ERTDKDSRLP LILVGNKSDL VEYSSMETIL PIMNQYTEIE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%. Binds to an epitope located within the peptide sequence VTSRWIPLIN as determined by overlapping synthetic peptides.
Mitochondrial GTPase involved in mitochondrial trafficking. RHOT1 is probably involved in control of anterograde transport of mitochondria and their subcellular distribution.
Specifications
RHOT1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXI2 | |
RHOT1 | |
Recombinant protein corresponding to Human RHOT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL1083 | |
Unconjugated | |
RHOT1 | |
2210403N23Rik; AA415293; AF244542; AI834919; Arht1; C430039G08Rik; LOW QUALITY PROTEIN: mitochondrial Rho GTPase 1; MGC128572 protein; Miro1; MIRO-1; mitochondrial Rho (MIRO) GTPase 1; mitochondrial Rho GTPase 1; rac-GTP binding protein-like protein; Rac-GTP-binding protein-like protein; ras homolog family member T1; Ras homolog gene family member T1; ras homolog gene family, member T1; Rhot1 | |
Mouse | |
Protein A | |
RUO | |
55288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only