Learn More
Risedronic acid, MedChemExpress
MedChemExpress Risedronic acid (Risedronate ) is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.
Brand: Medchem Express HY-B0148
This item is not returnable.
View return policy
Description
Risedronic acid (Risedronate ) is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.Specifications
Risedronic acid | |
105462-24-6 | |
500 mg | |
C7H11NO7P2 | |
0.1 M NaOH : 11 mg/mL (38.85 mM; ultrasonic and adjust pH to 7 with NaOH) ∣H2O : 0.67 mg/mL (2.37 mM; Need ultrasonic) | |
283.11 | |
98.0% |
Solid | |
White | |
Research | |
Risedronate | |
OC(P(O)(O)=O)(P(O)(O)=O)CC1=CN=CC=C1 | |
283.11 | |
Research |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Risedronic acid, MedChemExpress