Learn More
Risedronic acid, MedChemExpress
MedChemExpress Risedronic acid (Risedronate ) is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.
Brand:  Medchem Express HY-B0148
Encompass_Preferred
 View more versions of this product
Product Code. 16452046
  • 306.45 EUR / 500mg
Description
Risedronic acid (Risedronate ) is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.
Specifications
Specifications
Risedronic acid
105462-24-6
500 mg
C7H11NO7P2
0.1 M NaOH : 11 mg/mL (38.85 mM; ultrasonic and adjust pH to 7 with NaOH) ∣H2O : 0.67 mg/mL (2.37 mM; Need ultrasonic)
283.11
98.0%
Solid
White
Research
Risedronate
OC(P(O)(O)=O)(P(O)(O)=O)CC1=CN=CC=C1
283.11
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Risedronic acid, MedChemExpress