missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF32 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17430-25UL
This item is not returnable.
View return policy
Description
RNF32 Polyclonal antibody specifically detects RNF32 in Human samples. It is validated for ImmunofluorescenceSpecifications
RNF32 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
FKSG33, HSD15, ring finger protein 32 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KGHSSKKDNLAVNAVALQDHILHDLQLRNLSVADHSKTQVQKKENKSLKRDTKAIIDTGLKKTTQCPKL | |
25 μg | |
Zinc Finger | |
140545 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |