missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF32 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
360.68 EUR - 575.58 EUR
Specifications
Antigen | RNF32 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18384055
|
Novus Biologicals
NBP3-17430-25UL |
25 μg |
360.68 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331477
|
Bio-Techne
NBP3-17430-100UL |
100 μg |
575.58 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNF32 Polyclonal antibody specifically detects RNF32 in Human samples. It is validated for ImmunofluorescenceSpecifications
RNF32 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Zinc Finger | |
PBS, pH 7.2, 40% glycerol | |
140545 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
FKSG33, HSD15, ring finger protein 32 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KGHSSKKDNLAVNAVALQDHILHDLQLRNLSVADHSKTQVQKKENKSLKRDTKAIIDTGLKKTTQCPKL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |