missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Round Stainless Steel Pipette Boxes

Sterilize and store pipettes in a secure, stainless steel box. Bochem™ Round Stainless Steel Pipette Boxes are tube-shaped and include a silicone-covered bottom and lid.

Brand:  Bochem™ 8983

 View more versions of this product

Product Code. 10129550

  • 109.00 EUR / Each
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 18/10 stainless steel construction.
  • Silicone-covered bottom and lid.
  • Sterilizable up to 205°C.
  • Rotating the lid opens or closes the box.
Specifications

Specifications

18/10 Stainless Steel, Silicone
Glass pipets
450mm
65mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers