Learn More
Bochem™ Round Stainless Steel Pipette Boxes
Sterilize and store pipettes in a secure, stainless steel box. Bochem™ Round Stainless Steel Pipette Boxes are tube-shaped and include a silicone-covered bottom and lid.
Brand:  Bochem™ 8982
 View more versions of this product
Product Code. 10302901
  • 112.00 EUR / Each
Estimated Shipment: 17-10-2024
Description
  • 18/10 stainless steel construction.
  • Silicone-covered bottom and lid.
  • Sterilizable up to 205°C.
  • Rotating the lid opens or closes the box.
Specifications
Specifications
18/10 Stainless Steel, Silicone
Glass pipets
380mm
65mm