Learn More
Bochem™ Round Stainless Steel Pipette Boxes
Sterilize and store pipettes in a secure, stainless steel box. Bochem™ Round Stainless Steel Pipette Boxes are tube-shaped and include a silicone-covered bottom and lid.
Brand: Bochem™ 8984
This item is not returnable.
View return policy
Description
- 18/10 stainless steel construction.
- Silicone-covered bottom and lid.
- Sterilizable up to 205°C.
- Rotating the lid opens or closes the box.
Specifications
18/10 Stainless Steel, Silicone | |
Glass pipets | |
380mm | |
80mm |