missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16964-1-AP-150UL
This item is not returnable.
View return policy
Description
SET1A is a component of a histone methyltransferase.Specifications
RPS5 | |
Polyclonal | |
Unconjugated | |
RPS5 | |
40S ribosomal protein S5, ribosomal protein S5, RPS5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20103, 25538, 6193 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24050, P46782, P97461 | |
RPS5 | |
RPS5 Fusion Protein Ag10686 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |