missing translation for 'onlineSavingsMsg'
Learn More

RPS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 EUR - 410.00 EUR

Specifications

Antigen RPS5
Concentration 0.23 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16876764
View Documents
Proteintech
16964-1-AP-20UL
20 μL
145.00 EUR
20µL
Please to purchase this item. Need a web account? Register with us today!
16866764
View Documents
Proteintech
16964-1-AP-150UL
150 μL
410.00 EUR
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SET1A is a component of a histone methyltransferase.
Specifications

Specifications

RPS5
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P24050, P46782, P97461
20103, 25538, 6193
RPS5 Fusion Protein Ag10686
Primary
-20°C
RPS5
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
40S ribosomal protein S5, ribosomal protein S5, RPS5
RPS5
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications