missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5101376
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RPS7.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S7E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPS7 | |
Polyclonal | |
Unconjugated | |
RPS7 | |
40S ribosomal protein S7; DBA8; Mtu; ribosomal protein S7; RPS7; Rps7A; S7; S8; Small ribosomal subunit protein eS7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20115, 29258, 6201 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62081, P62082, P62083 | |
RPS7 | |
A synthesized peptide derived from human RPS7, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPS7 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction