missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAGA/RRAGB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5101819
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RRAGA/B.
Ras-homologous GTPases constitute a large family of signal transducers that alternate between an activated, GTP-binding state and an inactivated, GDP-binding state. These proteins represent cellular switches that are operated by GTP-exchange factors and factors that stimulate their intrinsic GTPase activity. All GTPases of the Ras superfamily have in common the presence of six conserved motifs involved in GTP/GDP binding, three of which are phosphate-/magnesium-binding sites (PM1-PM3) and three of which are guanine nucleotide-binding sites (G1-G3). Transcript variants encoding distinct isoforms have been identified. [provided by RefSeq, Jul 2008].
Specifications
RRAGA/RRAGB | |
Polyclonal | |
Unconjugated | |
RRAGB | |
1300010C19Rik; adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; AI255374; bA465E19.1; FIP1; FIP-1; GTP-binding protein ragB; rag A; Rag B; Raga; Ragb; Ras related GTP binding A; Ras related GTP binding B; Ras-related GTP binding A; Ras-related GTP binding B; ras-related GTP-binding protein A; Ras-related GTP-binding protein B; Ras-related GTP-binding protein ragA; RGD:619804}; RGD:619805}; RRAGA; rraga {ECO:0000312; RragB; rragB {ECO:0000312 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10325, 10670, 117043, 117044, 245670, 68441 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5VZM2, Q63486, Q63487, Q6NTA4, Q7L523, Q80X95 | |
RRAGA, RRAGB | |
A synthesized peptide derived from human RRAGA(Accession Q7L523), corresponding to amino acid residues N269-R313. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RRAGA/RRAGB Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction