Learn More
SCHOTT AG Lighting and Imaging™ RS 232 Computer Interface
RS 232 Computer interface (USB 1.1 optional) to set parameters via computer. To use with Multifunction Controller MC1500 (400 000).
Brand: SCHOTT AG Lighting and Imaging™ 400030
This item is not returnable.
View return policy
Description
- RS 232 Computer interface (USB 1.1 optional) to set parameters via computer.
- To use with Multifunction Controller MC1500 (400 000).
Specifications
Converter Cable | |
Stereo Microscopy Illumination | |
5 ft. |
RS 232 | |
Multifunction VisiLED Controller MC1500 (400 000) | |
1500 mm |
Product Content Correction
Product Title
SCHOTT AG Lighting and Imaging™ RS 232 Computer Interface > RS 232 Computer Interface; For Use With: Multifunction VisiLED Controller MC1500 (400 000)
Spot an opportunity for improvement?Share a Content Correction