Learn More
Fisherbrand™ Cork Stoppers
For use in education, homebrewing and arts and crafts industries.
Brand:  Fisherbrand™ CB030/S/Z
 View more versions of this product
Product Code. 12803834
Description
Description
Available in different sizes
Specifications
Specifications
32 mm
30 mm
Cork
10/Pk.
33 mm
Cork Stopper
Product Title
Fisherbrand™ Cork Stoppers > Base Diameter: 30mm