Learn More
(S)-(+)-3-Aminopiperidine dihydrochloride, 98%, Thermo Scientific Chemicals
41.00 EUR - 119.60 EUR
Chemical Identifiers
CAS | 334618-07-4 |
---|---|
Molecular Formula | C5H14Cl2N2 |
Molecular Weight (g/mol) | 173.08 |
MDL Number | MFCD03427036 |
InChI Key | GGPNYXIOFZLNKW-UHFFFAOYNA-N |
Synonym | s-3-aminopiperidine dihydrochloride, s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine dihydrochloride, s-3-aminopiperidine 2hcl, 3s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine 2hcl, 3-piperidinamine, dihydrochloride, 3s, 3s-3-piperidinamine dihydrochloride, 3s-3-aminopiperidine dihydrochloride, s-piperidin-3-ylamine dihydrochloride |
PubChem CID | 16218278 |
IUPAC Name | (3S)-piperidin-3-amine;dihydrochloride |
SMILES | Cl.Cl.NC1CCCNC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15457715
|
Thermo Scientific Alfa Aesar
H27700.MD |
250 mg | N/A | N/A | N/A | |||||
11312809
|
Thermo Scientific Alfa Aesar
H27700.03 |
1 g |
119.60 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
334618-07-4 | |
173.08 | |
GGPNYXIOFZLNKW-UHFFFAOYNA-N | |
16218278 | |
Cl.Cl.NC1CCCNC1 |
C5H14Cl2N2 | |
MFCD03427036 | |
s-3-aminopiperidine dihydrochloride, s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine dihydrochloride, s-3-aminopiperidine 2hcl, 3s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine 2hcl, 3-piperidinamine, dihydrochloride, 3s, 3s-3-piperidinamine dihydrochloride, 3s-3-aminopiperidine dihydrochloride, s-piperidin-3-ylamine dihydrochloride | |
(3S)-piperidin-3-amine;dihydrochloride |
Specifications
334618-07-4 | |
C5H14Cl2N2 | |
Hygroscopic | |
GGPNYXIOFZLNKW-UHFFFAOYNA-N | |
Cl.Cl.NC1CCCNC1 | |
173.08 | |
173.08 | |
(S)-(+)-3-Aminopiperidine dihydrochloride |
∽210°C (decomposition) | |
MFCD03427036 | |
s-3-aminopiperidine dihydrochloride, s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine dihydrochloride, s-3-aminopiperidine 2hcl, 3s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine 2hcl, 3-piperidinamine, dihydrochloride, 3s, 3s-3-piperidinamine dihydrochloride, 3s-3-aminopiperidine dihydrochloride, s-piperidin-3-ylamine dihydrochloride | |
+1.6° (c=1 in Water) | |
(3S)-piperidin-3-amine;dihydrochloride | |
16218278 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.