Learn More
Fisherbrand™ S-Series Heated Ultrasonic Cleaning Bath
With state of the art microprocessor controlled ultrasonic cleaning and sweep technology. Fisherbrand™ S-Series Ultrasonic Cleaners offer a comprehensive range of analogue ultrasonic water baths covering the requirements for laboratory research, sonochemistry and degassing applications.
Brand: Fisherbrand™ 1002279
This item is not returnable.
View return policy
Description
- Long lifespan from cavitation resistant stainless steel tanks
- LED display showing set and remaining time and pre-set and actual temperature
- Ergonomically shaped plastic carrying handles
- Newly developed cover reduces noise and accelerates the heating up process
- Easy draining of cleaning liquid though the drain duct via a dial on the side
- Operating panel arranged so that liquid cannot enter the electronics
- Degas function key
Specifications
V2A Stainless Steel | |
9.5 L | |
230VAC, 550w | |
Elmasonic S 100 (H) | |
5.9 kg |
230 VAC | |
278 x 365 x 264 mm | |
Laboratory, medical sector, dental laboratory, jewellery, watch sector, optician, workshop and service, industry | |
30°C to 80°C |
Videos
-
Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
-
Fisherbrand Ultrasonic baths S series
Fisherbrand Essentials - Ultrasonic cleaning baths
File size: 19.3 MB
Fisherbrand Operating manual S series ultrasonic baths
File size: 882 kB
ELM012 ELMA TEC CLEAN A2
File size: 25 kB
Product Content Correction
Product Title
Fisherbrand™ S-Series Heated Ultrasonic Cleaning Bath > Capacity: 9.5L; With Heating
Spot an opportunity for improvement?Share a Content Correction