Learn More
DuPont™ Tyvek 500 Xpert Hooded Coverall
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
Brand:  DuPont™ D15553614
896.74 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
 View more versions of this product
Product Code. 17412893
  • 1115.00 EUR / Pack of 100
Estimated Shipment: 05-06-2025
Description
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
  • Robust yet lightweight
  • 3-piece hood for optimal fit to head and face when turning
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Ample crotch area for freedom of movement
  • Large, easy-to-grasp zipper puller
  • Very low inward leakage thanks to optimized design
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5X-Large
Zipper
(EU) 2016/425, EN 14126, EN 1149-5
White
Stitched
Special Offers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
DuPont™ Tyvek 500 Xpert Hooded Coverall > Size: 4X-Large
The user should make the risk assessment to determine the protection required.