Learn More
Deutsch Neumann™ Safety Gas Hose
For gas burners DIN 30 665 and all gases according to DVGW G260.
Brand: Deutsch Neumann™ 3608300
This item is not returnable.
View return policy
Description
- System pressure up to 100 mbar
- Red-brown in color
- Manufactured from chloroprene (CR)
- Hardness 60 Shore A
- DVGW-tested and registered
Specifications
Chloroprene | |
Gas burners meeting DIN 30 665 | |
60 (Shore A) | |
Gas Safety Hose |
3000 mm | |
Red/Brown | |
100 m bar |
Product Content Correction
Product Title
Deutsch Neumann™ Safety Gas Hose > Safety Gas Hose
Spot an opportunity for improvement?Share a Content Correction