Learn More
Cytiva Sample Valve
Brand: Cytiva 29023896
Description
For use with the AKTA start automated protein purification system. The sample valve allows either the buffer or the sample to enter the flow path. The sample valve enables the direct application of the sample onto the column using the pump.
Specifications
Enables the direct application of the sample onto the column using the pump | |
AKTA start Automated Protein Purification System |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%