Learn More
SARS-CoV-2 N Protein Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814506S
This item is not returnable.
View return policy
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 N Protein Monoclonal antibody specifically detects SARS-CoV-2 N Protein in Virus samples. It is validated for ELISA, Western BlotSpecifications
SARS-CoV-2 N Protein | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P59595 | |
Mouse | |
Protein A/G | |
RUO | |
1489678 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
OTI4B9 | |
Unconjugated | |
N | |
COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid | |
Full length recombinant SARS-CoV-2 N protein produced in HEK293. | |
30 μL | |
Primary | |
Virus | |
Antibody | |
IgG1 |