missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 N Protein Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00 EUR - 540.87 EUR
Specifications
Antigen | SARS-CoV-2 N Protein |
---|---|
Clone | OTI4B9 |
Concentration | 1 mg/mL |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17529294
|
Origene Technologies
TA814506S |
30 μL |
242.00 EUR
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17509294
|
Origene Technologies
TA814506 |
100 μL |
540.87 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 N Protein Monoclonal antibody specifically detects SARS-CoV-2 N Protein in Virus samples. It is validated for ELISA, Western BlotSpecifications
SARS-CoV-2 N Protein | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid | |
Full length recombinant SARS-CoV-2 N protein produced in HEK293. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
N |
OTI4B9 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Virus | |
P59595 | |
1489678 | |
IgG1 | |
Protein A/G | |
Antibody |