Learn More
Fisherbrand Screw ISO97380 M4X12, 3 ea/bag (Transparent coverL)
Brand:  Fisherbrand 88861214
4.80 EUR valid until 2025-03-31
Use promo code "24162" to get your promotional price.
Screw ISO97380 M4X12, 3 ea/bag (Transparent coverL)
Product Code. 16258420
  • 5.45 EUR / Each
Estimated Shipment: 01-09-2025
Special Offers
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24162
    Expires2025-03-31
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Product Title
Fisherbrand Screw ISO97380 M4X12, 3 ea/bag (Transparent coverL) >