Learn More
3M™ SecureFit™ 100 Safety Glasses
Click to view available options
Model:
SF101AF-BLK-EU
SF101AF-BLU-EU
SF101AS-BLK-EU
SF101AS-BLU-EU
SF102AF-BLU-EU
SF103AF-BLU-EU
SF107AF-BLU-EU
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Description
- Economical robust 3M™ SecureFit™ Safety Glasses range
- 3M's innovative SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization. The temples flex and self-adjust to the size of wearer's head
- Slim wrap around design to look better on smaller faces
- Economical Bulk package (5-pack) available (black frame only)
- Flat and flexible temple design combines well with 3M™ Protective Earmuffs
- Wraparound design improves the security of fit and prevents slipping
- Easy to clean
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Certifications/Compliance (Europe) | EN 166:2001 |
Lens Tint | Indoor/Outdoor Light Gray |
Product Type | Protective Eyewear |
Frame Color | Blue |
Model | SF107AF-BLU-EU |
Frame Material | Polycarbonate |
UV Protection | UV400 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ SecureFit™ 100 Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction