Learn More
Brady™ Self-laminating Thermal Transfer Labels for M611, BMP61 and BMP71
Vinyl labels for use with Thermal Transfer Portable Printers.
Brand:  Brady™ 018432
 View more versions of this product
Product Code. 11548913
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Labels for the TLS2200™ and TLS PC Link™ Thermal Transfer Portable Printers
  • This material is UL approved with its respective ribbon
  • TLS2200™ parts can also be printed on BMP™71 Label Printer provided roll adapter (M71-ADAPT) is used
Specifications
Specifications
Not Autoclavable
White, Transparent
Self-laminating Vinyl
Non-sterile
BMP™61, BMP™71, BradyPrinter M611 Mobile Label Printers, TLS-PC LINK™ Thermal Labeling System
250 Labels
-70°C to +70°C
41.3 mm
38.1 mm
38.1 mm
Permanent Acrylic
Matte
Thermal Transfer
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling
1
Square
0.102 mm
10 AWG to 4 AWG
38.1 mm
12.7 mm