Learn More
Steris™ Self-Sealing sterilization Pouches
Provides an effective means of sterilization, safe handling and storage of all items until the moment they are used. Steris Corporation™ Self-Sealing sterilization Pouches are CE marked for steam and EO gas.
47.50 EUR - 97.90 EUR
Specifications
Certifications/Compliance | CE Marked |
---|---|
For Use With (Equipment) | Steam and Gas EO Sterilizers |
Quantity | 200/Pk. |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
15448782
|
Steris™
NWKP2035 |
200 x 350mm |
97.90 EUR
Pack of 200 |
Estimated Shipment: 05-05-2025
Log in to see stock. |
||||||
15428782
|
Steris™
NWKP0927 |
90 x 270mm |
47.50 EUR
Pack of 200 |
Estimated Shipment: 05-05-2025
Log in to see stock. |
||||||
Description
- Self-sealing bags eliminate the need for sealing devices
- Excellent adhesive strength resistant to steam exposure
- Non-toxic water-based indicator inks
- Perforated flap for easy closing
Specifications
CE Marked | |
Steam and Gas EO Sterilizers | |
200/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Steris™ Self-Sealing sterilization Pouches