Learn More
Fisherbrand™ Septum for 20mm Crimp Cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 20 10 2451
Description
- 20mm Septa
Specifications
Freeze Drying Stopper | |
20 mm | |
1000 |
Gray | |
Butyl | |
Freeze Drying Stopper |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Fisherbrand™ Septum for 20mm Crimp Cap > Butyl grey,42° shore A
Spot an opportunity for improvement?Share a Content Correction