Learn More
Greiner Bio-One VACUETTE™ Z Serum Blood Collection Tubes
VACUETTE™ Z Serum Blood Collection Tubes are used in determinations for clinical chemistry, immunology and serology. Available in Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator. The inner wall of the tube is coated with silica particles which activate clotting when the tubes are turned.
222.86 EUR
Specifications
Additive Z Serum Clot Activator
Sterility Sterile
For Use With (Equipment) VACUETTE™ Blood Collection Needles
View More Specs
Products 2
Product Code Brand Capacity (Metric) Closure Color Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Color Price Quantity & Availability  
12532936
View Documents
Greiner Bio-One
455092
9 mL Red with Black Ring
222.86 EUR
Pack of 1200
13068662
View Documents
Greiner Bio-One
454096
2 mL Red with White Ring
222.86 EUR
Pack of 1200
Description
Description
VACUETTE™ Z Serum Blood Collection Tubes are used in determinations for clinical chemistry, immunology and serology. Available in Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator. The inner wall of the tube is coated with silica particles which activate clotting when the tubes are turned.
  • Tubes are supplied in packs of 50 pieces
  • For venus blood collection
  • Primarily used for drawing blood for clinical chemistry, immunology and serology testing
  • Specially coated with microscopic silica particles, which activate the coagulation process
  • Made out of virtually unbreakable PET plastic
  • Provides improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Available in: Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator
  • CE marked
Specifications
Specifications
Z Serum Clot Activator
Sterile
VACUETTE™ Blood Collection Needles
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Greiner Bio-One VACUETTE™ Z Serum Blood Collection Tubes