Learn More
SF3B3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand:  Invitrogen PA518075
338.68 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Product Code. 11366043
  • 508.00 EUR / 100µg
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
This gene encodes subunit 3 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex transcription coactivator-HAT complex, and the TFTC -containing complex). These complexes may function in chromatin modification, transcription, splicing, and DNA repair.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Title
SF3B3 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated