Learn More
Trajan™ Glass Lined Bulkhead Unions with Locknut
Glass lined stainless steel construcion. Bulkhead unions with locknut
Brand: Trajan™ 1236306
This item is not returnable.
View return policy
Description
- Allows plate mounting of union
- For use with fused silica
- Low dead volume
- High temperature applications
Specifications
Bulkhead Union | |
No restriction | |
Glass-lined Stainless Steel | |
0.8 mm |