Learn More
Trajan™ Glass Lined Bulkhead Unions with Locknut
Glass lined stainless steel construcion. Bulkhead unions with locknut
229.00 EUR
Product Code | Brand | Diameter (Metric) Inner | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Description | Price | Quantity & Availability | ||||
15239163
|
Trajan™
1236306 |
0.8 mm | No restriction |
229.00 EUR
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||
15219163
|
Trajan™
1236300 |
0.5 mm | 0.35 mm restriction (at one end) |
229.00 EUR
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||
15229163
|
Trajan™
1236302 |
0.4 mm | 0.25 mm restriction (at one end) |
229.00 EUR
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||
Description
- Allows plate mounting of union
- For use with fused silica
- Low dead volume
- High temperature applications
Specifications
Bulkhead Union | |
Glass-lined Stainless Steel |