missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe Sharpsbin
GREENER_CHOICE

A cost-effective solution for lowering the CO2 emissions. Eco-friendly sharps and hazardous waste containers, produced from renewable resources using green energy. Reducing CO2 emissions by up to 66%, documented in a full LCA. Available in sizes from 0.5 L to 50 L.

Brand:  Woodsafe 1005120

 View more versions of this product

Product Code. 18737338

  • 144.06 EUR / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Alert:

Follow instructions for safe and correct use


Description

Description

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry.
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be temporarily closed to conceal the contents when not in use and securely permanently closed for transportation to the final disposal.
  • Natural colour and smell from wood - No migration of substances - EN - tested for toys and food
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic
Specifications

Specifications

0.5 L
Temporary and permanent closure
Round
0.132 gal.
8.1 x 8.1 x 14.4 cm (3.19 x 3.19 x 5.67 in.)
50
Yes , when empty
Non-sterile
Bin + Lid + Label
Cardboard
Biohazard Waste Container (Benchtop)
EN ISO 23907-1:2019. UN3291, ADR Approved
Brown
Environmental friendly Sharpsbin
Brown, Yellow
Single-use
Round
8.1 x 8.1 x 14.4 cm
Autoclavable
Safe Disposal of sharps and potentially infectious waste or medical waste
Biocomposite
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers