missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe Sharpsbin
GREENER_CHOICE

A cost-effective solution for lowering the CO2 emissions. Eco-friendly sharps and hazardous waste containers, produced from renewable resources using green energy. Reducing CO2 emissions by up to 66%, documented in a full LCA. Available in sizes from 0.5 L to 50 L.

144.06 EUR - 291.56 EUR

Specifications

Certifications/Compliance EN ISO 23907-1:2019. UN3291, ADR Approved
Closure Color Brown
Description Environmental friendly Sharpsbin
Color Brown, Yellow
Disposable Single-use
View More Specs

Alert:

Follow instructions for safe and correct use


Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
18737338
GREENER_CHOICE
View accessory products View Documents
Woodsafe
1005120
0.5 L
144.06 EUR
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
18727338
GREENER_CHOICE
View accessory products View Documents
Woodsafe
1020120
2.3 L
291.56 EUR
Pack of 70
Please to purchase this item. Need a web account? Register with us today!
18772815
GREENER_CHOICE
View accessory products View Documents
Woodsafe
1060110
6 L
226.00 EUR
Pack of 30
Please to purchase this item. Need a web account? Register with us today!
18762825
GREENER_CHOICE
View accessory products View Documents
Woodsafe
1120110
12 L
213.16 EUR
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry.
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be temporarily closed to conceal the contents when not in use and securely permanently closed for transportation to the final disposal.
  • Natural colour and smell from wood - No migration of substances - EN - tested for toys and food
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic
Specifications

Specifications

EN ISO 23907-1:2019. UN3291, ADR Approved
Environmental friendly Sharpsbin
Single-use
Autoclavable
Cardboard
Biohazard Waste Container (Benchtop)
Brown
Brown, Yellow
Non-sterile
Bin + Lid + Label
Biocomposite
Videos
SDS
Documents

Documents

Product Certifications