Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule.
Brand:  Thermo Scientific™ 290MA202
78.77 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10670616
  • 79.16 EUR / Pack of 10
Estimated Shipment: 14-05-2025
Description
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal
  • expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Ferrule
All SilTite Kits
Pk. of 10 Ferrules
290 MA202
SilTite Metal
0.32 mm
Product Title
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals > Replacement Ferrule; 0.32mm; For Use With Agilent