Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule.
Brand: Thermo Scientific™ 290MT222
78.77 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
- SilTite metal ferrules and nuts are made from the same material and have the same thermal
- expansion coefficient
- Base of the ferrule is flat and forms a perfect seal with the MS interface
- The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
- Unlike other ferrules, do not need re-tightening after installation
Specifications
Replacement SilTite Ferrules | |
Thermo Scientific DSQ GC-MS Interface Kits | |
Pk. of 10 Ferrules |
290 MT222 | |
SilTite Metal | |
0.32 mm |