missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This volumetric solution was checked by means of potentiometric methods using Scharlau's disodium oxalate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).
- Molecular Formula: AgNO3
- Molar Mass: 169.87g/mol
- CAS: 7761-88-8
- EIN ECS No.: 231-853-9
- Factor: 0.999–1.001
Specifications
Specifications
CAS | 7761-88-8 |
Physical Form | Solution |
Quantity | 1 L |
Molecular Formula | AgNO3 |
MDL Number | MFCD00003414 |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
SMILES | [Ag+].[O-][N+]([O-])=O |
IUPAC Name | silver(1+) nitrate |
Molecular Weight (g/mol) | 169.87 |
Show More |
Safety and Handling

Product Identifier
- Silver nitrate, solution 0,1 mol/l (0,1 N)
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P362-Take off contaminated clothing.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Silver nitrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Silver Nitrate, 0.1M/l Solution, Scharlab >
Spot an opportunity for improvement?Share a Content Correction