Learn More
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™ Product Code.: 10640423
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™
Product Code. 10640423
10 L, Bag in box
Click to view available options
Quantity:
2.5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Product Code. 10640423
Brand: Fisher Chemical J/7330/24
Request Bulk or Custom Format
AgNO3, CAS Number-7761-88-8, silver nitrate, lunar caustic, argerol, silver mononitrate, silvernitrate, silver i nitrate, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, 2.5L, CHEBI:32130, Colorless, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate, 3414, 169.872
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerolShow MoreShow Less
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O
Specifications
Color Colorless
Packaging Bag in box
Quantity 10 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Silver nitrate, 0.1M solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    22
    Hours
    19
    Minutes
    29
    Seconds
    30
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
    Days
    53
    Hours
    19
    Minutes
    29
    Seconds
    30
Product Content Correction
Product Title
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™ >