Learn More
VITLAB™ Simplex™ Dispenser
Brand:  VITLAB™ 1601504
Product Code. 10201692
Specifications
≤±0.5%
Bottletop Dispenser
DIN EN 285, DE-M marked
0.5 to 5 mL
Product Title
VITLAB™ Simplex™ Dispenser > Capacity: 0.5 to 5mL; Accuracy: ≤±0.5%