Learn More
SMAD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 511500
334.68 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
SMAD3 Polyclonal antibody specifically detects SMAD3 in Human samples. It is validated for ChIP Assay, ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.
Specifications
SMAD3 | |
Polyclonal | |
Unconjugated | |
Smad3 | |
AU022421; DKFZp586N0721; DKFZp686J10186; hMAD-3; hSMAD3; HSPC193; HsT17436; JV15-2; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 3; MAD homolog 3a; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MAD-3; madh3; madh3a; madh3-A; MGC60396; mMad3; mothers against decapentaplegic homolog 3; mothers against decapentaplegic homolog 3a; mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD; Smad 3; SMAD family member 3; SMAD family member 3 L homeolog; SMAD family member 3a; SMAD, mothers against DPP homolog 3; Smad3; smad3.L; smad3a; TGF beta response effector Smad3; TGF-beta response effector Smad3; transcription factor; transcription factor Smad3; wu:fa99e03; XELAEV_18018454mg; XenMLP; Xmad3; XSmad3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4088 | |
-20°C | |
Liquid |
ChIP Assay, ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P84022 | |
Smad3 | |
A 20 amino acid synthetic peptide derived from a central portion of the linker domain of human Smad3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.