Learn More
Fisherbrand™ Snap Caps
Made from chemically resistant virgin plastic, Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange. They are ideal for use in science, research, and other laboratories.
Brand: Fisherbrand™ BC16WHITE
Description
Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs. Suitable for all standard test tube sizes, they fit rimless and plastic tubes with an outside diameter of 13mm, 16mm, 19mm, 25mm and 38mm. The three flexible fins molded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur. Bacti-Caps™ are made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters.- Improved culture growth
- Color coded
- Controlled gas interchange
Specifications
Snap Cap | |
White |
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Fisherbrand™ Snap Caps > White
Spot an opportunity for improvement?Share a Content Correction