missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
2 kg
Description
- It is used as a buffer in the buffering range of pH 3.6-5.6.
- It is also used in the purification and precipitation of nucleic acids, protein crystallization, Staining of gels in protein gel electrophoresis, HPLC, mordant dyeing, DNA microarray studies of E. coli response, investigation of protein unfolding during reverse phase chromatography.
- It is used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents.
- It is also used as substrate for acetokinase. Sodium acetate is the sodium salt of acetic acid.

Specifications
Specifications
Boiling Point | >400°C with decomposition (literature) |
Melting Point | 324°C with decomposition (literature) |
Color | White |
pH | 7.0 to 9.2 (5% aq soln) |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Beilstein | 3595639 |
Flash Point | >250°C / >482°F (lit.) |
Solubility Information | Soluble freely in water; moderately soluble in alcohol. |
Grade | ACS |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium acetate, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction