missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Arsenite 0.05 M For Laboratory Use and Titrations, Chem Lab
Brand: Chem Lab CL05.1402.1000
This item is not returnable.
View return policy
Specifications
Colorless | |
100°C | |
0.05 M | |
129.91 | |
1 L |
7784-46-5 | |
0°C | |
1.01 g/mL | |
Polyethylene | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium arsenite 0.05 mol/l
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Arsenic Trioxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Arsenite 0.05 M For Laboratory Use and Titrations, Chem Lab > Sodium Arsenite 0.05 M For Laboratory Use and Titrations, 1 L
Spot an opportunity for improvement?Share a Content Correction