Learn More
Sodium Arsenite Solution, 0.05M, Fisher Chemical™ Product Code.: 12933604
compliance-icons
Sodium Arsenite Solution, 0.05M, Fisher Chemical™
Product Code. 12933604
1000 mL
Click to view available options
Quantity:
1000 mL
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 12933604
Brand: Fisher Chemical VL 286 1000
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 1000 mL
Concentration 0.05 mol/L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1065832 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 286 1000
Lot Number1001329 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 286 1000
Lot Number1015381 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 286 1000
Lot Number1053950 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 286 1000
Lot Number1055522 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 286 1000
compliance-icons
Product Identifier
  • Sodium arsenite 0.05M Volumetric Solution (VL 286)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Product Content Correction
Product Title
Sodium Arsenite Solution, 0.05M, Fisher Chemical™ >