missing translation for 'onlineSavingsMsg'
Learn More
Sodium carbonate anhydrous, for analysis, ExpertQ, Scharlab Product Code.: 11388534

Sodium carbonate anhydrous, for analysis, ExpertQ, Scharlab

Product Code. 11388534
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11388534

Brand: Scharlab SO01161000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 497-19-8
Quantity 1 kg
Molecular Formula CNa2O3
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Molecular Weight (g/mol) 105.99
Grade ExpertQ
compliance-icons
Product Identifier
  • Sodium carbonate anhydrous, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium carbonate anhydrous, for analysis, ExpertQ, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.