missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Description
Sodium chloride is used in a wide variety of biochemical applications, including:
- Intravenous fluids (normal saline is 0.9% w/v in water10)
- Density gradients
- A diluent to increase ionic strength in buffers or culture media
- In salt-out procedures in the isolation of DNA
- In high concentrations for preservation of foods, since bacteria cannot grow in high salt conditions
A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as &minus’21°C, depending on the rate of stirring and the size of the ice chunks. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.

Specifications
Specifications
Physical Form | Crystalline Powder |
Quantity | 10 kg |
Solubility Information | Soluble in water (357mg/mL at 25°C; 384mg/mL at 100°C), glycerol (100mg/mL); very slightly soluble in ethanol. Almost insoluble in concentrated HCl. |
IUPAC Name | sodium chloride |
Formula Weight | 58.44g/mol |
Percent Purity | ≥99.5% |
Grade | Molecular Biology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction