Learn More
Sodium hexanitritocobaltate(III), Thermo Scientific Chemicals
50.55 EUR - 182.91 EUR
Chemical Identifiers
CAS | 14649-73-1 |
---|---|
Molecular Formula | CoN6Na3O12 |
Molecular Weight (g/mol) | 403.933 |
MDL Number | MFCD00003512 |
InChI Key | ZEBNDJQGEZXBCR-UHFFFAOYSA-H |
Synonym | cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11 |
PubChem CID | 159748 |
IUPAC Name | trisodium;cobalt(3+);hexanitrite |
SMILES | N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473534
|
Thermo Scientific Alfa Aesar
A11216.18 |
50 g |
50.55 EUR
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483534
|
Thermo Scientific Alfa Aesar
A11216.30 |
250 g |
182.91 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with acids, reducing agents and amines.
Chemical Identifiers
14649-73-1 | |
403.933 | |
ZEBNDJQGEZXBCR-UHFFFAOYSA-H | |
159748 | |
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3] |
CoN6Na3O12 | |
MFCD00003512 | |
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11 | |
trisodium;cobalt(3+);hexanitrite |
Specifications
220°C (decomposition) | |
CoN6Na3O12 | |
MFCD00003512 | |
14,8603 | |
Soluble in water. | |
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3] | |
403.933 | |
403.94 | |
Sodium hexanitritocobaltate (III) |
14649-73-1 | |
Na3Co(NO2)6 | |
UN1479 | |
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11 | |
ZEBNDJQGEZXBCR-UHFFFAOYSA-H | |
trisodium;cobalt(3+);hexanitrite | |
159748 | |
Odorless |
Safety and Handling
GHS H Statement
H272-H334-H351-H335-H315-H319-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P210-P220-P221-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P370+P378q-P501c
H272-H315-H317-H319-H334-H335-H350
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
missing translation for 'einecsNumber' : 237-077-7
missing translation for 'rtecsNumber' : GF9480000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium hexanitrocobaltate(III)
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contains : Sodium hexanitrocobaltate(III)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.