missing translation for 'onlineSavingsMsg'
Learn More
Sodium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals Product Code.: 11343747

Sodium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals

Product Code. 11343747
20 L
Click to view available options
Quantity:
1 L
4 L
20 L
This item is not returnable. View return policy

Product Code. 11343747

Brand: Thermo Scientific Alfa Aesar 035625.M4

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium hydroxide, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 20 L
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Solubility Miscible with glycerol, ethanol and methanol; Immiscible with acetone and ether
Formula Weight 40
Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.4%; Water: 99.6%
Chemical Name or Material Sodium hydroxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP16L520 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number035625.M4,035625.K2,035625.K7
Lot NumberN26L520 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number035625.K2,035625.M4,035625.K7
Lot NumberN26L519 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number035625.K2,035625.M4,035625.K7
Lot NumberZ14K505 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number035625.K2,035625.M4,035625.K7
Lot NumberZ14K504 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number035625.K2,035625.M4,035625.K7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.