missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.
Specifications
Specifications
pH | >13 |
Color | Colorless |
Boiling Point | 100°C |
CAS | 1310-73-2, 7732-18-5 |
Molecular Formula | HNaO |
Concentration | 2M |
Density | 1.09g/mL |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Solubility Information | Soluble in Water |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling

Product Identifier
- Sodium hydroxide 2 mol/l
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : sodium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide Solution, 2M, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction