missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Pellets, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
1 kg
Description
- Synonyms: Caustic soda
- Molecular Formula: NaOH
- Molar Mass: 40.00g/mol
- CAS: 1310-73-2
- EIN ECS No.: 215-185-5
- Carbonates (as Na2CO3): max. 1.0%
- Silicates (SiO2): max. 0.001%
- Ammonium Hydroxide Precipitate: max. 0.02%
Specifications
Specifications
Boiling Point | 1390°C |
CAS | 1310-73-2 |
Melting Point | 323°C |
Molecular Formula | HNaO |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Solubility Information | Soluble in water at 20°C |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Quantity | 1 kg |
SMILES | [OH-].[Na+] |
IUPAC Name | sodium;hydroxide |
Show More |
Safety and Handling

Product Identifier
- Sodium hydroxide, pellets, reagent grade, ACS, ISO, Reag. Ph Eur
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide Pellets, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction