Learn More
Sodium Hydroxide Solution, 0.111M (0.111N), for analysis of milk, Fisher Chemical™
Click to view available options
Quantity:
1 L
5 L
Specifications
Specifications
Quantity | 1 L |
Normality | 0.111 |
Packaging | HDPE Plastic Bottle |
Traceability to NIST | Traceable to NIST, used to titrate the lactic acid present in milk |
Physical Form | Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Product Title
Sodium Hydroxide Solution, 0.111M (0.111N), for analysis of milk, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction