missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide solution 0.5mol/L, Fisher Chemical™ Product Code.: 12970764

Sodium Hydroxide solution 0.5mol/L, Fisher Chemical™

Product Code. 12970764
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 12970764

Brand: Fisher Chemical SOL 123 100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 1310-73-2, 7732-18-5
Packaging Bottle
Quantity 100 mL
Molecular Formula HNaO
Concentration 0.5 M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
PubChem CID 14798
ChEBI CHEBI:32145
Concentration or Composition (by Analyte or Components) 0.5mol/L
Chemical Name or Material Sodium Hydroxide solution
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide 5M Volumetric Solution (SOL 123)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide solution 0.5mol/L, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.